Learn More
4-Chloro-3-(trifluoromethyl)phenyl isocyanate, 98%, Thermo Scientific Chemicals
992.85 SEK - 2555.05 SEK
Chemical Identifiers
CAS | 327-78-6 |
---|---|
Molecular Formula | C8H3ClF3NO |
Molecular Weight (g/mol) | 221.56 |
MDL Number | MFCD00013874 |
InChI Key | NBJZEUQTGLSUOB-UHFFFAOYSA-N |
Synonym | 4-chloro-3-trifluoromethyl phenyl isocyanate, 1-chloro-4-isocyanato-2-trifluoromethyl benzene, 4-chloro-3-trifluoromethyl phenylisocyanate, benzene, 1-chloro-4-isocyanato-2-trifluoromethyl, 4-chloro-3-trifluoromethylphenyl isocyanate, 4-chloro-3-trifluoromethoxy phenyl isocyanate, 1-chloro-4-isocyanato-2-trifluoromethyl-benzene, 4-chloro-3-trifluoromethyl benzenisocyanate, 3-trifluoromethyl-4-chloro-phenyl isocyanate, pubchem5027 |
PubChem CID | 2733265 |
IUPAC Name | 1-chloro-4-isocyanato-2-(trifluoromethyl)benzene |
SMILES | C1=CC(=C(C=C1N=C=O)C(F)(F)F)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10503231
|
Thermo Scientific Acros
309130050 |
5 g | Glass bottle |
992.85 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15720859
|
Thermo Scientific Acros
309130250 |
25 g | Glass bottle |
2555.05 SEK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
327-78-6 | |
221.56 | |
NBJZEUQTGLSUOB-UHFFFAOYSA-N | |
2733265 | |
C1=CC(=C(C=C1N=C=O)C(F)(F)F)Cl |
C8H3ClF3NO | |
MFCD00013874 | |
4-chloro-3-trifluoromethyl phenyl isocyanate, 1-chloro-4-isocyanato-2-trifluoromethyl benzene, 4-chloro-3-trifluoromethyl phenylisocyanate, benzene, 1-chloro-4-isocyanato-2-trifluoromethyl, 4-chloro-3-trifluoromethylphenyl isocyanate, 4-chloro-3-trifluoromethoxy phenyl isocyanate, 1-chloro-4-isocyanato-2-trifluoromethyl-benzene, 4-chloro-3-trifluoromethyl benzenisocyanate, 3-trifluoromethyl-4-chloro-phenyl isocyanate, pubchem5027 | |
1-chloro-4-isocyanato-2-(trifluoromethyl)benzene |
Specifications
327-78-6 | |
100.0 | |
White to Yellow | |
Authentic | |
C8H3ClF3NO | |
MFCD00013874 | |
NBJZEUQTGLSUOB-UHFFFAOYSA-N | |
1-chloro-4-isocyanato-2-(trifluoromethyl)benzene | |
2733265 | |
98% | |
4-Chloro-3-(trifluoromethyl)phenyl isocyanate |
97.5 | |
29.0°C to 37.0°C | |
99°C | |
97.5% min. (GC) | |
ClC6H3(CF3)NCO | |
4-chloro-3-trifluoromethyl phenyl isocyanate, 1-chloro-4-isocyanato-2-trifluoromethyl benzene, 4-chloro-3-trifluoromethyl phenylisocyanate, benzene, 1-chloro-4-isocyanato-2-trifluoromethyl, 4-chloro-3-trifluoromethylphenyl isocyanate, 4-chloro-3-trifluoromethoxy phenyl isocyanate, 1-chloro-4-isocyanato-2-trifluoromethyl-benzene, 4-chloro-3-trifluoromethyl benzenisocyanate, 3-trifluoromethyl-4-chloro-phenyl isocyanate, pubchem5027 | |
C1=CC(=C(C=C1N=C=O)C(F)(F)F)Cl | |
221.56 | |
221.56 | |
Low Melting Solid, Powder or Chunks |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.
Harmful if swallowed.
Fatal if inhaled.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Warning
RUO – Research Use Only