missing translation for 'onlineSavingsMsg'
Learn More
4-Cyclohexylaniline, 97%, Thermo Scientific Chemicals Product Code.: 11332056

4-Cyclohexylaniline, 97%, Thermo Scientific Chemicals

Product Code. 11332056
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11332056

Brand: Thermo Scientific Alfa Aesar L03184.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

4-Cyclohexylaniline is used as a starting material for the preparation of 1-cyclohexyl-4-nitroso-benzene. It is used as a reactant in nucleophilic acylation to form N-(4-cyclohexylphenyl)acetamide by reacting with acetyl halides. It is also involved in the synthesis of N-(4-cyclohexylphenyl)-2-morpholino-acetamide by reaction with 2-morpholinoacetyl fluoride. Further, it reacts with 4-bromo-4-oxo-butanoic acid to get 4-[(4-cyclohexylphenyl)amino]-4-oxo-butanoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyclohexylaniline is used as a starting material for the preparation of 1-cyclohexyl-4-nitroso-benzene. It is used as a reactant in nucleophilic acylation to form N-(4-cyclohexylphenyl)acetamide by reacting with acetyl halides. It is also involved in the synthesis of N-(4-cyclohexylphenyl)-2-morpholino-acetamide by reaction with 2-morpholinoacetyl fluoride. Further, it reacts with 4-bromo-4-oxo-butanoic acid to get 4-[(4-cyclohexylphenyl)amino]-4-oxo-butanoic acid.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6373-50-8
Molecular Formula C16H12Cl2N4OS
Molecular Weight (g/mol) 379.26
MDL Number MFCD00001454
InChI Key YDTTUXLKBBQRHA-UHFFFAOYSA-N
Synonym benzenamine, 4-cyclohexyl, 4-cyclohexylbenzenamine, aniline, p-cyclohexyl, p-cyclohexylaniline, 4-cyclohexylphenyl amine, 4-cyclohexylphenylamine, 4-cyclohexylanilin, 4-cyclohexyl-aniline, acmc-20aivf, p-cyclohexylphenylamine
PubChem CID 80764
IUPAC Name 4-cyclohexylaniline
SMILES CC1=CC=C(C=C1)C1=NN=C(NC(=O)NC2=CC=C(Cl)C=C2Cl)S1

Specifications

Melting Point 52°C to 56°C
Boiling Point 166°C (13 mmHg)
Quantity 5 g
Beilstein 2965799
Solubility Information Insoluble in water.
Formula Weight 175.28
Percent Purity 97%
Chemical Name or Material 4-Cyclohexylaniline

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10251913 Certificate TypeCertificate of Analysis Date08/08/2024 Catalog NumberL03184.03,L03184.06
Lot Number10241396 Certificate TypeCertificate of Analysis Date22/06/2022 Catalog NumberL03184.03,L03184.06
Lot Number10148966 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL03184.03,L03184.06
Lot Number10151915 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL03184.03,L03184.06
Lot Number10189714 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL03184.03,L03184.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Cyclohexylaniline, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.