missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxy-3-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific™

218.00 SEK - 975.00 SEK

Chemical Identifiers

CAS 252901-50-1
Molecular Formula C10H9F3O3
Molecular Weight (g/mol) 234.17
MDL Number MFCD13196578
InChI Key KWEQYUCUGSDCHR-UHFFFAOYSA-N
Synonym 4-ethoxy-3-trifluoromethyl benzoic acid, benzoic acid, 4-ethoxy-3-trifluoromethyl
PubChem CID 23508153
IUPAC Name 4-ethoxy-3-(trifluoromethyl)benzoic acid
SMILES CCOC1=C(C=C(C=C1)C(O)=O)C(F)(F)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15426957
SDS View Documents
Thermo Scientific Alfa Aesar
H34048.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15436957
SDS View Documents
Thermo Scientific Alfa Aesar
H34048.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

252901-50-1
234.17
KWEQYUCUGSDCHR-UHFFFAOYSA-N
23508153
CCOC1=C(C=C(C=C1)C(O)=O)C(F)(F)F
C10H9F3O3
MFCD13196578
4-ethoxy-3-trifluoromethyl benzoic acid, benzoic acid, 4-ethoxy-3-trifluoromethyl
4-ethoxy-3-(trifluoromethyl)benzoic acid

Specifications

252901-50-1
MFCD13196578
KWEQYUCUGSDCHR-UHFFFAOYSA-N
4-ethoxy-3-(trifluoromethyl)benzoic acid
23508153
97%
C10H9F3O3
4-ethoxy-3-trifluoromethyl benzoic acid, benzoic acid, 4-ethoxy-3-trifluoromethyl
CCOC1=C(C=C(C=C1)C(O)=O)C(F)(F)F
234.17
234.17
4-Ethoxy-3-(trifluoromethyl)benzoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.