missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-(trifluoromethyl)benzaldehyde, 98%, Thermo Scientific™

619.00 SEK - 2030.00 SEK

Chemical Identifiers

CAS 90176-80-0
Molecular Formula C8H4F4O
Molecular Weight (g/mol) 192.113
MDL Number MFCD00061313
InChI Key NONOHEMDNFTKCZ-UHFFFAOYSA-N
Synonym 4-fluoro-2-trifluoromethyl benzaldehyde, 2-trifluoromethyl-4-fluorobenzaldehyde, 4-fluoro-2-trifluoromethyl-benzaldehyde, benzaldehyde, 4-fluoro-2-trifluoromethyl, 5-fluoro-2-formylbenzotrifluoride, alpha,alpha,alpha,4-tetrafluoro-o-tolualdehyde, pubchem4241, ksc487q2t, paragos 530044
PubChem CID 522940
IUPAC Name 4-fluoro-2-(trifluoromethyl)benzaldehyde
SMILES C1=CC(=C(C=C1F)C(F)(F)F)C=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11429047
SDS View Documents
Thermo Scientific Alfa Aesar
B20621.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11439047
SDS View Documents
Thermo Scientific Alfa Aesar
B20621.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALDEHYDES, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Fluoro-2-(trifluoromethyl)benzaldehyde, 98%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.