missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoro-3-(tetrazol-5-yl)phenylboronic acid, 95%, Thermo Scientific™
2531.98 SEK - 8976.00 SEK
Chemical Identifiers
CAS | 1009303-56-3 |
---|---|
Molecular Formula | C7H6BFN4O2 |
Molecular Weight (g/mol) | 207.96 |
MDL Number | MFCD13176533 |
InChI Key | FELFJHZMQYCKPT-UHFFFAOYSA-N |
Synonym | 4-fluoro-3-tetrazol-5-yl phenylboronic acid, 4-fluoro-3-1h-1,2,3,4-tetrazol-5-yl phenylboronic acid, 4-fluoro-3-1h-tetrazol-5-yl phenyl boronic acid, 4-fluoro-3-1h-tetrazol-5-yl phenylboronic acid |
PubChem CID | 11287236 |
IUPAC Name | [4-fluoro-3-(2H-tetrazol-5-yl)phenyl]boronic acid |
SMILES | OB(O)C1=CC(C2=NNN=N2)=C(F)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11956751
|
Thermo Scientific Acros
445460010 |
1 g |
2531.98 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11966751
|
Thermo Scientific Acros
445460050 |
5 g | N/A | ||||||
Description
Chemical Identifiers
1009303-56-3 | |
207.96 | |
FELFJHZMQYCKPT-UHFFFAOYSA-N | |
11287236 | |
OB(O)C1=CC(C2=NNN=N2)=C(F)C=C1 |
C7H6BFN4O2 | |
MFCD13176533 | |
4-fluoro-3-tetrazol-5-yl phenylboronic acid, 4-fluoro-3-1h-1,2,3,4-tetrazol-5-yl phenylboronic acid, 4-fluoro-3-1h-tetrazol-5-yl phenyl boronic acid, 4-fluoro-3-1h-tetrazol-5-yl phenylboronic acid | |
[4-fluoro-3-(2H-tetrazol-5-yl)phenyl]boronic acid |
Specifications
1009303-56-3 | |
Glass Bottle | |
MFCD13176533 | |
FELFJHZMQYCKPT-UHFFFAOYSA-N | |
[4-fluoro-3-(2H-tetrazol-5-yl)phenyl]boronic acid | |
11287236 | |
95% |
94% min. (HPLC) | |
C7H6BFN4O2 | |
4-fluoro-3-tetrazol-5-yl phenylboronic acid, 4-fluoro-3-1h-1,2,3,4-tetrazol-5-yl phenylboronic acid, 4-fluoro-3-1h-tetrazol-5-yl phenyl boronic acid, 4-fluoro-3-1h-tetrazol-5-yl phenylboronic acid | |
OB(O)C1=CC(C2=NNN=N2)=C(F)C=C1 | |
207.96 | |
207.96 | |
4-Fluoro-3-(tetrazol-5-yl)phenylboronic acid |
Safety and Handling
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 4-Fluoro-3-(tetrazol-5-yl)phenylboronic acid
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
4-Fluoro-3-(tetrazol-5-yl)phenylboronic acid, 95%, Thermo Scientific™