missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-4'-methoxybenzophenone, 98%, Thermo Scientific™

Product Code. 15438248
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 345-89-1
Molecular Formula C14H11FO2
Molecular Weight (g/mol) 230.24
MDL Number MFCD00055469
InChI Key VWGWRNBIAWTWIB-UHFFFAOYSA-N
Synonym 4-fluoro-4'-methoxybenzophenone, 4-fluorophenyl 4-methoxyphenyl methanone, 4-fluorophenyl-4-methoxyphenyl methanone, 4-fluorphenyl 4-methoxyphenyl methanon, pubchem3309, acmc-209i8i, 4-methoxy-4'-fluorobenzophenone, 4-fluoro-4-methoxybenzophenone, 4-fluoro-4'-methoxy-benzophenone, 4-fluorophenyl 4-methoxyphenyl ketone
PubChem CID 67664
IUPAC Name (4-fluorophenyl)-(4-methoxyphenyl)methanone
SMILES COC1=CC=C(C=C1)C(=O)C1=CC=C(F)C=C1

Specifications

Color White
Quantity 1 g
Formula Weight 230.23
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-4'-methoxybenzophenone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.