missing translation for 'onlineSavingsMsg'
Learn More
4-Fluorophenylacetyl chloride, 97%, Thermo Scientific Chemicals Product Code.: 11314106

4-Fluorophenylacetyl chloride, 97%, Thermo Scientific Chemicals

Product Code. 11314106
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11314106

Brand: Thermo Scientific Alfa Aesar L19235.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

4-Fluorophenylacetyl chloride is used as an acylating agent in organic synthesis. It is used to prepare monoacylated pyrazolidine by reacting with pyrazolidine. It is also used in the preparation of N-acyl isothiouronium chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorophenylacetyl chloride is used as an acylating agent in organic synthesis. It is used to prepare monoacylated pyrazolidine by reacting with pyrazolidine. It is also used in the preparation of N-acyl isothiouronium chloride.

Solubility
Miscible with dichloromethane.

Notes
Moisture Sensitive. Store in a cool place. Incompatible with water, alcohols, oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 459-04-1
Molecular Formula C8H6ClFO
Molecular Weight (g/mol) 172.58
MDL Number MFCD00674181
InChI Key SIOJFYRPBYGHOO-UHFFFAOYSA-N
Synonym 4-fluorophenylacetyl chloride, 2-4-fluorophenyl acetyl chloride, 4-fluorophenylacetylchloride, 4-fluorobenzeneacetyl chloride, p-fluorophenylacetyl chloride, benzeneacetyl chloride, 4-fluoro, 2-4-fluorophenyl ethanoyl chloride, 4-fluoro phenylacetyl chloride, pubchem17517, acmc-1aivq
PubChem CID 3744988
IUPAC Name 2-(4-fluorophenyl)acetyl chloride
SMILES FC1=CC=C(CC(Cl)=O)C=C1

Specifications

Density 1.259
Boiling Point 60°C (8 mmHg)
Flash Point 107°C (224°F)
Refractive Index 1.512
Quantity 5 g
UN Number UN3265
Beilstein 972148
Sensitivity Moisture sensitive
Solubility Information Miscible with dichloromethane.
Formula Weight 172.59
Percent Purity 97%
Chemical Name or Material 4-Fluorophenylacetyl chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10193876 Certificate TypeCertificate of Analysis Date09/02/2023 Catalog NumberL19235.06,L19235.14
Lot NumberC16S003 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL19235.06,L19235.14
Lot Number10180564 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL19235.06,L19235.14
Lot NumberB07S001 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL19235.06,L19235.14
Lot NumberK08S010 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL19235.06,L19235.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Fluorophenylacetyl chloride, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.