Learn More
4-Hydroxy-6-methyl-2-(trifluoromethyl)quinoline, 97%, Thermo Scientific™
372.00 SEK - 1192.84 SEK
Chemical Identifiers
CAS | 1701-20-8 |
---|---|
Molecular Formula | C11H8F3NO |
Molecular Weight (g/mol) | 227.186 |
MDL Number | MFCD00153192 |
InChI Key | UNVMZLUVACVTDT-UHFFFAOYSA-N |
Synonym | 4-hydroxy-6-methyl-2-trifluoromethyl quinoline, 6-methyl-2-trifluoromethyl quinolin-4-ol, 6-methyl-2-trifluoromethyl-1h-quinolin-4-one, 2-trifluoromethyl-6-methylquinolin-4-ol, 6-methyl-2-trifluoromethylquinolin-4-ol, 4-quinolinol, 6-methyl-2-trifluoromethyl, 4-hydroxy-6-methyl-2-trifluoromethyl-quinoline, 6-methyl-2-trifluoromethyl-4-quinolinol, 6-methyl-2-trifluoromethyl quinolin-4 1h-one, maybridge1_004906 |
PubChem CID | 2775088 |
IUPAC Name | 6-methyl-2-(trifluoromethyl)-1H-quinolin-4-one |
SMILES | CC1=CC2=C(C=C1)NC(=CC2=O)C(F)(F)F |
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
11449137
|
Thermo Scientific Alfa Aesar
B20675.03 |
1 g | N/A | ||||||
11459137
|
Thermo Scientific Alfa Aesar
B20675.06 |
5 g | N/A | ||||||
Beschreibung
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemische Identifikatoren
1701-20-8 | |
227.186 | |
UNVMZLUVACVTDT-UHFFFAOYSA-N | |
2775088 | |
CC1=CC2=C(C=C1)NC(=CC2=O)C(F)(F)F |
C11H8F3NO | |
MFCD00153192 | |
4-hydroxy-6-methyl-2-trifluoromethyl quinoline, 6-methyl-2-trifluoromethyl quinolin-4-ol, 6-methyl-2-trifluoromethyl-1h-quinolin-4-one, 2-trifluoromethyl-6-methylquinolin-4-ol, 6-methyl-2-trifluoromethylquinolin-4-ol, 4-quinolinol, 6-methyl-2-trifluoromethyl, 4-hydroxy-6-methyl-2-trifluoromethyl-quinoline, 6-methyl-2-trifluoromethyl-4-quinolinol, 6-methyl-2-trifluoromethyl quinolin-4 1h-one, maybridge1_004906 | |
6-methyl-2-(trifluoromethyl)-1H-quinolin-4-one |
Spezifikation
1701-20-8 | |
C11H8F3NO | |
4-hydroxy-6-methyl-2-trifluoromethyl quinoline, 6-methyl-2-trifluoromethyl quinolin-4-ol, 6-methyl-2-trifluoromethyl-1h-quinolin-4-one, 2-trifluoromethyl-6-methylquinolin-4-ol, 6-methyl-2-trifluoromethylquinolin-4-ol, 4-quinolinol, 6-methyl-2-trifluoromethyl, 4-hydroxy-6-methyl-2-trifluoromethyl-quinoline, 6-methyl-2-trifluoromethyl-4-quinolinol, 6-methyl-2-trifluoromethyl quinolin-4 1h-one, maybridge1_004906 | |
CC1=CC2=C(C=C1)NC(=CC2=O)C(F)(F)F | |
227.186 | |
227.19 | |
4-Hydroxy-6-methyl-2-(trifluoromethyl)quinoline |
252°C to 253°C | |
MFCD00153192 | |
UNVMZLUVACVTDT-UHFFFAOYSA-N | |
6-methyl-2-(trifluoromethyl)-1H-quinolin-4-one | |
2775088 | |
97% |
Sicherheit und Handhabung
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.