missing translation for 'onlineSavingsMsg'
Learn More
4-Hydroxyphthalic acid, 98%, Thermo Scientific Chemicals Product Code.: 11477397

4-Hydroxyphthalic acid, 98%, Thermo Scientific Chemicals

Product Code. 11477397
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11477397

Brand: Thermo Scientific Alfa Aesar A15091.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

4-Hydroxyphthalic acid is used as a precursor to prepare 3,5-dichloro-4-hydroxyphthalic acid using the reagent aqueous acetic acid and chlorine. Further, it is also used as a starting material in the manufacture of N-(4-carboxyphenyl)-4-acetoxyphthalimide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxyphthalic acid is used as a precursor to prepare 3,5-dichloro-4-hydroxyphthalic acid using the reagent aqueous acetic acid and chlorine. Further, it is also used as a starting material in the manufacture of N-(4-carboxyphenyl)-4-acetoxyphthalimide.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 610-35-5
Molecular Formula C8H6O5
Molecular Weight (g/mol) 182.131
MDL Number MFCD00013984
InChI Key MWRVRCAFWBBXTL-UHFFFAOYSA-N
Synonym 1,2-benzenedicarboxylic acid, 4-hydroxy, 4-hydroxybenzene-1,2-dicarboxylic acid, 4-hydroxy-1,2-benzenedicarboxylic acid, 3,4-dicarboxyphenol, 4-hydroxy-phthalic acid, acmc-1avol, ksc357m7h, 4-hydroxyphthalic acid, 4-hydroxy-1,2-benzenedicarboxylicacid, 1,2-benzenedicarboxylicacid, 4-hydroxy
PubChem CID 11881
ChEBI CHEBI:27600
IUPAC Name 4-hydroxyphthalic acid
SMILES C1=CC(=C(C=C1O)C(=O)O)C(=O)O

Specifications

Melting Point ∼190°C (decomposition)
Quantity 25 g
Solubility Information Slightly miscible with water.
Formula Weight 182.13
Percent Purity 98%
Chemical Name or Material 4-Hydroxyphthalic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10184375 Certificate TypeCertificate of Analysis Date16/01/2023 Catalog NumberA15091.06,A15091.14,A15091.22
Lot Number10243828 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog NumberA15091.06,A15091.14,A15091.22
Lot NumberJ9312A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15091.06,A15091.14,A15091.22
Lot Number10138116 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15091.06,A15091.14,A15091.22
Lot Number10160601 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15091.06,A15091.14,A15091.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Hydroxyphthalic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.