missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-2-methylbenzaldehyde, 97%, Thermo Scientific Chemicals

Product Code. 10526694
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52289-54-0
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD02261771
InChI Key WICYVKGMEJSDAO-UHFFFAOYSA-N
Synonym 2-methyl-4-methoxybenzaldehyde, 4-methoxy-2-methyl-benzaldehyde, benzaldehyde, 4-methoxy-2-methyl, 2-methyl-p-anisaldehyde, 2-methyl-anisaldehyde, acmc-1aoyo, 4-methoxy-2-methylbenzaldehyd, 4-methoxy-2-methylbenzaldehyde, 3-furane-2-amido benzoic acid
PubChem CID 283285
IUPAC Name 4-methoxy-2-methylbenzaldehyde
SMILES CC1=C(C=CC(=C1)OC)C=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1070g/mL
Boiling Point 262°C to 267°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH30C6H3(CH3)CHO
Quantity 5 g
Specific Gravity 1.107
Formula Weight 150.18
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Methoxy-2-methylbenzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methoxy-2-methylbenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H371-May cause damage to organs.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'