missing translation for 'onlineSavingsMsg'
Learn More
4-Methoxy-4'-nitrobenzophenone, 97%, Thermo Scientific Chemicals Product Code.: 11415377

4-Methoxy-4'-nitrobenzophenone, 97%, Thermo Scientific Chemicals

Product Code. 11415377
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11415377

Brand: Thermo Scientific Alfa Aesar B22720.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

4-Methoxy-4'-nitrobenzophenone is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxy-4′-nitrobenzophenone is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1151-94-6
Molecular Formula C14H11NO4
Molecular Weight (g/mol) 257.25
MDL Number MFCD00178348
InChI Key DXVSAELNVPXMSQ-UHFFFAOYSA-N
Synonym 4-methoxy-4'-nitrobenzophenone, 4-methoxyphenyl 4-nitrophenyl methanone, 4-methoxyphenyl-4-nitrophenyl methanone, methanone, 4-methoxyphenyl 4-nitrophenyl, 4-methoxy-phenyl-4-nitro-phenyl-methanone, acmc-1buw2, 4-methoxy-4-nitrobenzophenone, 4-methoxy-4?-nitrobenzophenone, 4-methoxy-4/'-nitrobenzophenoneShow MoreShow Less
PubChem CID 347772
IUPAC Name (4-methoxyphenyl)-(4-nitrophenyl)methanone
SMILES COC1=CC=C(C=C1)C(=O)C1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 125°C to 127°C
Quantity 25 g
Formula Weight 257.25
Percent Purity 97%
Chemical Name or Material 4-Methoxy-4'-nitrobenzophenone

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberJ5731A Certificate TypeCertificate of Analysis Date01/05/2023 Catalog NumberB22720.03,B22720.14,B22720.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Methoxy-4'-nitrobenzophenone, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.