Learn More
4-Methoxy-N-(2-methylbenzyl)benzamide, 97%, Thermo Scientific™
2060.00 SEK - 6168.00 SEK
Chemical Identifiers
CAS | 331638-66-5 |
---|---|
Molecular Formula | C16H17NO2 |
Molecular Weight (g/mol) | 255.317 |
MDL Number | MFCD01154205 |
InChI Key | CVDCGDPYWBZBAN-UHFFFAOYSA-N |
Synonym | 4-methoxy-n-2-methylbenzyl benzamide, 4-methoxy-n-2-methylphenyl methyl benzamide, cbmicro_011456, cambridge id 6073509, 4-hydroxymethyl-n-2-methylbenzyl benzamide |
PubChem CID | 789734 |
IUPAC Name | 4-methoxy-N-[(2-methylphenyl)methyl]benzamide |
SMILES | CC1=CC=CC=C1CNC(=O)C2=CC=C(C=C2)OC |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
331638-66-5 | |
255.317 | |
CVDCGDPYWBZBAN-UHFFFAOYSA-N | |
789734 | |
CC1=CC=CC=C1CNC(=O)C2=CC=C(C=C2)OC |
C16H17NO2 | |
MFCD01154205 | |
4-methoxy-n-2-methylbenzyl benzamide, 4-methoxy-n-2-methylphenyl methyl benzamide, cbmicro_011456, cambridge id 6073509, 4-hydroxymethyl-n-2-methylbenzyl benzamide | |
4-methoxy-N-[(2-methylphenyl)methyl]benzamide |
Specifications
331638-66-5 | |
MFCD01154205 | |
CVDCGDPYWBZBAN-UHFFFAOYSA-N | |
4-methoxy-N-[(2-methylphenyl)methyl]benzamide | |
789734 | |
97% |
C16H17NO2 | |
4-methoxy-n-2-methylbenzyl benzamide, 4-methoxy-n-2-methylphenyl methyl benzamide, cbmicro_011456, cambridge id 6073509, 4-hydroxymethyl-n-2-methylbenzyl benzamide | |
CC1=CC=CC=C1CNC(=O)C2=CC=C(C=C2)OC | |
255.317 | |
274.31 | |
4-Methoxy-N-(2-methylbenzyl)benzamide |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.