missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methyl-3-heptanol, erythro + threo, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in ethanol, ether, acetone. Slightly soluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Soluble in ethanol, ether, acetone. Slightly soluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Specifications
Specifications
Density | 0.83 |
Boiling Point | 98°C to 99°C (75 mmHg) |
Flash Point | 59°C (138°F) |
Refractive Index | 1.431 |
Quantity | 1 g |
UN Number | UN1987 |
Beilstein | 1733008 |
Solubility Information | Soluble in ethanol,ether,acetone. Slightly soluble in water. |
Formula Weight | 130.23 |
Percent Purity | 99% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'