missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3-(trifluoromethyl)aniline, 98+%, Thermo Scientific™

Product Code. 11411007
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65934-74-9
Molecular Formula C8H8F3N
Molecular Weight (g/mol) 175.154
MDL Number MFCD01631582
InChI Key JBCDCYFEJQHTTA-UHFFFAOYSA-N
Synonym 4-methyl-3-trifluoromethyl aniline, 5-amino-2-methylbenzotrifluoride, 3-trifluoromethyl-4-methylaniline, benzenamine, 4-methyl-3-trifluoromethyl, 4-methyl-5-trifluoromethyl aniline, 4-methyl-3-trifluoromethyl-phenylamine, 4-methyl-5-trifluoromethylaniline, 3-trifluoromethyl-p-toluidine, 4-methyl-3-trifluoromethyl phenylamine
PubChem CID 2737715
IUPAC Name 4-methyl-3-(trifluoromethyl)aniline
SMILES CC1=C(C=C(C=C1)N)C(F)(F)F

Specifications

Density 1.24
Boiling Point 204°C
Flash Point 102°C (216°F)
Refractive Index 1.4905
Quantity 5 g
UN Number UN2810
Beilstein 6776230
Formula Weight 175.16
Percent Purity ≥98%
Chemical Name or Material 4-Methyl-3-(trifluoromethyl)aniline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Methyl-3-(trifluoromethyl)aniline, 98+%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.