Learn More
4-(Methylsulfonyl)phenylacetic acid, 97%, Thermo Scientific Chemicals
589.32 SEK - 1834.23 SEK
Chemical Identifiers
CAS | 90536-66-6 |
---|---|
Molecular Formula | C9H10O4S |
Molecular Weight (g/mol) | 214.235 |
MDL Number | MFCD00216495 |
InChI Key | HGGWOSYNRVOQJH-UHFFFAOYSA-N |
Synonym | 4-methylsulfonyl phenylacetic acid, 4-methylsulphonylphenylacetic acid, 2-4-methylsulfonyl phenyl acetic acid, 4-mesylphenylacetic acid, 4-methanesulfonylphenyl acetic acid, 4-methylsulfonyl phenyl acetic acid, 4-methanesulfonyl phenylacetic acid, 2-4-methanesulfonylphenyl acetic acid, 4-methylsulfonylphenylacetic acid |
PubChem CID | 572345 |
IUPAC Name | 2-(4-methylsulfonylphenyl)acetic acid |
SMILES | CS(=O)(=O)C1=CC=C(C=C1)CC(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11318866
|
Thermo Scientific Alfa Aesar
L19504.06 |
5 g |
589.32 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11328866
|
Thermo Scientific Alfa Aesar
L19504.14 |
25 g |
1834.23 SEK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4-(Methylsulfonyl)phenylacetic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-(Methylsulfonyl)phenylacetic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Chemical Identifiers
90536-66-6 | |
214.235 | |
HGGWOSYNRVOQJH-UHFFFAOYSA-N | |
572345 | |
CS(=O)(=O)C1=CC=C(C=C1)CC(=O)O |
C9H10O4S | |
MFCD00216495 | |
4-methylsulfonyl phenylacetic acid, 4-methylsulphonylphenylacetic acid, 2-4-methylsulfonyl phenyl acetic acid, 4-mesylphenylacetic acid, 4-methanesulfonylphenyl acetic acid, 4-methylsulfonyl phenyl acetic acid, 4-methanesulfonyl phenylacetic acid, 2-4-methanesulfonylphenyl acetic acid, 4-methylsulfonylphenylacetic acid | |
2-(4-methylsulfonylphenyl)acetic acid |
Specifications
90536-66-6 | |
C9H10O4S | |
2646649 | |
Slightly soluble in water. | |
CS(=O)(=O)C1=CC=C(C=C1)CC(=O)O | |
214.235 | |
214.24 | |
4-(Methylsulfonyl)phenylacetic acid |
134°C to 138°C | |
MFCD00216495 | |
4-methylsulfonyl phenylacetic acid, 4-methylsulphonylphenylacetic acid, 2-4-methylsulfonyl phenyl acetic acid, 4-mesylphenylacetic acid, 4-methanesulfonylphenyl acetic acid, 4-methylsulfonyl phenyl acetic acid, 4-methanesulfonyl phenylacetic acid, 2-4-methanesulfonylphenyl acetic acid, 4-methylsulfonylphenylacetic acid | |
HGGWOSYNRVOQJH-UHFFFAOYSA-N | |
2-(4-methylsulfonylphenyl)acetic acid | |
572345 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only