missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferyl-2-Acetamido-2-Deoxy-?-D-Glucopyranoside, MP Biomedicals™

Product Code. 11452422
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
Questo articolo non è restituibile. Consulta la politica di reso
Questo articolo non è restituibile. Consulta la politica di reso

Substrate for the fluorimetric, phosphorimetric and spectrophotometric assay of N-acetyl-β-D-glucosaminidase and β-N-acetyl-D-hexosaminidase.

TRUSTED_SUSTAINABILITY

Specifica

CAS 37067-30-4
Melting Point 212°C to 222°C
Molecular Formula C18H21NO8
MDL Number MFCD00065481
Quantity 250 mg
Synonym 4-mu-glcnac, 4-methylumbelliferyl-n-acetyl-beta-d-glucosaminide, n-2s,3r,4r,5s,6r-4,5-dihydroxy-6-hydroxymethyl-2-4-methyl-2-oxo-2h-chromen-7-yl oxy tetrahydro-2h-pyran-3-yl acetamide, 4-methylumbelliferyl-2-acetamido-2-deoxy-beta-d-glucopyranoside, 4-methylumbelliferyl n-acetyl-, a-d-glucosaminide, 4-methylumbelliferyl n-acetyl-beta-d-glucosaminide, n-2s,3r,4r,5s,6r-4,5-dihydroxy-6-hydroxymethyl-2-4-methyl-2-oxochromen-7-yl oxy oxan-3-yl acetamide, glcnac-b-4mu, 4-methylumbelliferyl 2-acetamido-2-deoxy-, a-d-glucopyranoside, 4-methylumbelliferyl 2-acetamido-2-deoxy-b-d-glucopyranoside
Solubility Information Soluble in methanol, warm water (100mg/65ml) and DMF (20mg/mL)
InChI Key QCTHLCFVVACBSA-XWEABGILSA-N
SMILES CC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)OC1OC1=CC=C2C(C)=CC(=O)OC2=C1
IUPAC Name N-[(3R,4R,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-2-[(4-methyl-2-oxo-2H-chromen-7-yl)oxy]oxan-3-yl]acetamide
Molecular Weight (g/mol) 379.37
PubChem CID 2733787
Formula Weight 379.4
Physical Form Powder
Vedi altri risultati Mostra meno risultati
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'