Learn More
4-N-BOC-2-oxo-piperazine, 97%, Thermo Scientific Chemicals
480.18 SEK - 1649.90 SEK
Chemical Identifiers
CAS | 76003-29-7 |
---|---|
Molecular Formula | C9H16N2O3 |
Molecular Weight (g/mol) | 200.24 |
MDL Number | MFCD02181069 |
InChI Key | FCMLWBBLOASUSO-UHFFFAOYSA-N |
Synonym | 1-boc-3-oxopiperazine, 4-n-boc-2-oxo-piperazine, 4-boc-piperazinone, n-boc-3-oxopiperazine, 3-oxo-piperazine-1-carboxylic acid tert-butyl ester, tert-butyl 3-oxo-1-piperazinecarboxylate, 4-n-boc-piperazin-2-one, n-boc-piperazin-3-one, 1-piperazinecarboxylic acid, 3-oxo-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-3-oxopiperazine |
PubChem CID | 3157178 |
IUPAC Name | tert-butyl 3-oxopiperazine-1-carboxylate |
SMILES | CC(C)(C)OC(=O)N1CCNC(=O)C1 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
76003-29-7 | |
200.24 | |
FCMLWBBLOASUSO-UHFFFAOYSA-N | |
3157178 | |
CC(C)(C)OC(=O)N1CCNC(=O)C1 |
C9H16N2O3 | |
MFCD02181069 | |
1-boc-3-oxopiperazine, 4-n-boc-2-oxo-piperazine, 4-boc-piperazinone, n-boc-3-oxopiperazine, 3-oxo-piperazine-1-carboxylic acid tert-butyl ester, tert-butyl 3-oxo-1-piperazinecarboxylate, 4-n-boc-piperazin-2-one, n-boc-piperazin-3-one, 1-piperazinecarboxylic acid, 3-oxo-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-3-oxopiperazine | |
tert-butyl 3-oxopiperazine-1-carboxylate |
Specifications
76003-29-7 | |
100.0 | |
White | |
96% min. (GC) | |
MFCD02181069 | |
FCMLWBBLOASUSO-UHFFFAOYSA-N | |
tert-butyl 3-oxopiperazine-1-carboxylate | |
3157178 | |
97% | |
4-N-BOC-2-Oxo-piperazine |
96.0 | |
160.0°C to 163.0°C | |
Authentic | |
C9H16N2O3 | |
1-boc-3-oxopiperazine, 4-n-boc-2-oxo-piperazine, 4-boc-piperazinone, n-boc-3-oxopiperazine, 3-oxo-piperazine-1-carboxylic acid tert-butyl ester, tert-butyl 3-oxo-1-piperazinecarboxylate, 4-n-boc-piperazin-2-one, n-boc-piperazin-3-one, 1-piperazinecarboxylic acid, 3-oxo-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-3-oxopiperazine | |
CC(C)(C)OC(=O)N1CCNC(=O)C1 | |
200.24 | |
200.24 | |
Crystalline Powder |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
- 4-N-BOC-2-Oxo-piperazine
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.