Learn More
4-n-Pentylphenol, 98%, Thermo Scientific Chemicals
619.00 SEK - 6828.00 SEK
Chemical Identifiers
CAS | 14938-35-3 |
---|---|
Molecular Formula | C11H16O |
Molecular Weight (g/mol) | 164.25 |
MDL Number | MFCD00020211 |
InChI Key | ZNPSUQQXTRRSBM-UHFFFAOYSA-N |
Synonym | 4-n-pentylphenol, 4-amylphenol, phenol, 4-pentyl, 4-n-amylphenol, p-pentylphenol, p-amylphenol, phenol, p-pentyl, 4-pentyl-phenol, amyl p-hydroxybenzene, p-n-amylphenolShow More |
PubChem CID | 26975 |
ChEBI | CHEBI:34441 |
IUPAC Name | 4-pentylphenol |
SMILES | CCCCCC1=CC=C(O)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11484503
|
Thermo Scientific Alfa Aesar
A11514.06 |
5 g |
619.00 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11494503
|
Thermo Scientific Alfa Aesar
A11514.14 |
25 g |
2090.00 SEK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11404513
|
Thermo Scientific Alfa Aesar
A11514.22 |
100 g | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14938-35-3 | |
164.25 | |
ZNPSUQQXTRRSBM-UHFFFAOYSA-N | |
26975 | |
4-pentylphenol |
C11H16O | |
MFCD00020211 | |
4-n-pentylphenol, 4-amylphenol, phenol, 4-pentyl, 4-n-amylphenol, p-pentylphenol, p-amylphenol, phenol, p-pentyl, 4-pentyl-phenol, amyl p-hydroxybenzene, p-n-amylphenolShow More | |
CHEBI:34441 | |
CCCCCC1=CC=C(O)C=C1 |
Specifications
14938-35-3 | |
0.96 | |
133°C (271°F) | |
1.5145 | |
UN2430 | |
4-n-pentylphenol, 4-amylphenol, phenol, 4-pentyl, 4-n-amylphenol, p-pentylphenol, p-amylphenol, phenol, p-pentyl, 4-pentyl-phenol, amyl p-hydroxybenzene, p-n-amylphenol | |
CCCCCC1=CC=C(O)C=C1 | |
164.25 | |
CHEBI:34441 | |
98% |
24°C | |
342°C | |
C11H16O | |
MFCD00020211 | |
1364424 | |
ZNPSUQQXTRRSBM-UHFFFAOYSA-N | |
4-pentylphenol | |
26975 | |
164.25 | |
4-n-Pentylphenol |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALKYLPHENOLS, SOLID, N.O.S.
missing translation for 'einecsNumber' : 239-015-4
missing translation for 'rtecsNumber' : SM6750000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.