missing translation for 'onlineSavingsMsg'
Learn More

4-Nonylphenol, 99%, mixture of isomers, Thermo Scientific Chemicals

1940.74 SEK - 7230.43 SEK

Chemical Identifiers

CAS 84852-15-3
MDL Number MFCD00002396
InChI Key SNQQPOLDUKLAAF-UHFFFAOYSA-N
Synonym phenol, 2-nonyl, nonyl phenol, o-nonylphenol, nonyl, phenol,nonyl, 2-n-nonyl-phenol, ortho-n-nonyl-phenol, acmc-209t6o, bidd:er0079, nonylphenol, mixture of isomers
PubChem CID 67296
IUPAC Name 2-nonylphenol
SMILES CCCCCCCCCC1=CC=CC=C1O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10762022
View Documents Promotion Details
Thermo Scientific Acros
416240010
1 kg Glass Bottle
1940.74 SEK
1kg
Please to purchase this item. Need a web account? Register with us today!
10165363
View Documents Promotion Details
Thermo Scientific Acros
416240050
5 kg Plastic drum
7230.43 SEK
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

84852-15-3
SNQQPOLDUKLAAF-UHFFFAOYSA-N
67296
CCCCCCCCCC1=CC=CC=C1O
MFCD00002396
phenol, 2-nonyl, nonyl phenol, o-nonylphenol, nonyl, phenol,nonyl, 2-n-nonyl-phenol, ortho-n-nonyl-phenol, acmc-209t6o, bidd:er0079, nonylphenol, mixture of isomers
2-nonylphenol

Specifications

84852-15-3
100.0
293.0°C to 297.0°C
Authentic
C15H24O
MFCD00002396
0.94
Solubility in water: practically insoluble. Other solubilities: practically insoluble in diluted aq. naoh,soluble in benzene,chlorinated solvents,aniline,,alchols,ethylene glycol
CCCCCCCCCC1=CC=CC=C1O
220.35
220.35
Viscous Liquid
98.5
0.9400g/mL
141°C
98.5% min. (GC)
1.5105 to 1.5165
14, 6681
phenol, 2-nonyl, nonyl phenol, o-nonylphenol, nonyl, phenol,nonyl, 2-n-nonyl-phenol, ortho-n-nonyl-phenol, acmc-209t6o, bidd:er0079, nonylphenol, mixture of isomers
SNQQPOLDUKLAAF-UHFFFAOYSA-N
2-nonylphenol
67296
99%
4-Nonylphenol, Mixture of isomers
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Suspected of damaging fertility.
Suspected of damaging the unborn child.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 284-325-5

Product Identifier
  • 4-Nonylphenol, mixture of isomers

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only