Learn More
4-(Pentafluorothio)phenylacetic acid, 97%, Thermo Scientific™
534.00 SEK - 1613.00 SEK
Chemical Identifiers
CAS | 1839048-23-5 |
---|---|
Molecular Formula | C8H7F5O2S |
Molecular Weight (g/mol) | 262.194 |
MDL Number | MFCD16652426 |
InChI Key | HAIVYRSOFXWOFO-UHFFFAOYSA-N |
Synonym | 4-pentafluorosulfur phenylacetic acid, 4-pentafluoro-??-sulfanyl phenyl acetic acid, 4-pentafluorothio phenylacetic acid, 2-4-pentafluoro-lambda6-sulfanyl phenyl acetic acid |
PubChem CID | 66523467 |
IUPAC Name | 2-[4-(pentafluoro-$l^{6}-sulfanyl)phenyl]acetic acid |
SMILES | C1=CC(=CC=C1CC(=O)O)S(F)(F)(F)(F)F |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications4-(Pentafluorothio)phenylacetic acid participate in the synthesis and some transformations of allyl polyfluoroaryl sulfides and -sulfones.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
Chemical Identifiers
1839048-23-5 | |
262.194 | |
HAIVYRSOFXWOFO-UHFFFAOYSA-N | |
66523467 | |
C1=CC(=CC=C1CC(=O)O)S(F)(F)(F)(F)F |
C8H7F5O2S | |
MFCD16652426 | |
4-pentafluorosulfur phenylacetic acid, 4-pentafluoro-??-sulfanyl phenyl acetic acid, 4-pentafluorothio phenylacetic acid, 2-4-pentafluoro-lambda6-sulfanyl phenyl acetic acid | |
2-[4-(pentafluoro-$l^{6}-sulfanyl)phenyl]acetic acid |
Specifications
1839048-23-5 | |
MFCD16652426 | |
HAIVYRSOFXWOFO-UHFFFAOYSA-N | |
2-[4-(pentafluoro-$l^{6}-sulfanyl)phenyl]acetic acid | |
66523467 | |
97% |
C8H7F5O2S | |
4-pentafluorosulfur phenylacetic acid, 4-pentafluoro-??-sulfanyl phenyl acetic acid, 4-pentafluorothio phenylacetic acid, 2-4-pentafluoro-lambda6-sulfanyl phenyl acetic acid | |
C1=CC(=CC=C1CC(=O)O)S(F)(F)(F)(F)F | |
262.194 | |
262.19 | |
4-(Pentafluorothio)phenylacetic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.