missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Phenyl-5-(trifluoromethyl)thiophene-2-carboxylic acid, 95%, Thermo Scientific™
548.13 SEK - 4732.34 SEK
Chemical Identifiers
CAS | 208108-76-3 |
---|---|
Molecular Formula | C12H7F3O2S |
Molecular Weight (g/mol) | 272.241 |
InChI Key | GIUAGKGTDDIMHB-UHFFFAOYSA-N |
Synonym | 4-phenyl-5-trifluoromethyl thiophene-2-carboxylic acid, 4-phenyl-5-trifluoromethyl-2-thiophenecarboxylic acid, 2-thiophenecarboxylic acid, 4-phenyl-5-trifluoromethyl, 2-thiophenecarboxylicacid, 4-phenyl-5-trifluoromethyl, maybridge3_006560, 4-phenyl-5-trifluoromethyl-thiophene-2-carboxylic acid, 4-phenyl-5-trifluoromethyl-2-thiophene carboxylic acid |
PubChem CID | 2783372 |
IUPAC Name | 4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylic acid |
SMILES | C1=CC=C(C=C1)C2=C(SC(=C2)C(=O)O)C(F)(F)F |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10149602
|
Thermo Scientific Maybridge
SEW02660CB |
250 mg | Amber Glass Bottle |
548.13 SEK
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10644603
|
Thermo Scientific Maybridge
SEW02660DA |
1 g | Amber Glass Bottle |
1475.31 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10215334
|
Thermo Scientific Maybridge
SEW02660DE |
5 g | Amber Glass Bottle |
4732.34 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
208108-76-3 | |
272.241 | |
4-phenyl-5-trifluoromethyl thiophene-2-carboxylic acid, 4-phenyl-5-trifluoromethyl-2-thiophenecarboxylic acid, 2-thiophenecarboxylic acid, 4-phenyl-5-trifluoromethyl, 2-thiophenecarboxylicacid, 4-phenyl-5-trifluoromethyl, maybridge3_006560, 4-phenyl-5-trifluoromethyl-thiophene-2-carboxylic acid, 4-phenyl-5-trifluoromethyl-2-thiophene carboxylic acid | |
4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylic acid |
C12H7F3O2S | |
GIUAGKGTDDIMHB-UHFFFAOYSA-N | |
2783372 | |
C1=CC=C(C=C1)C2=C(SC(=C2)C(=O)O)C(F)(F)F |
Specifications
208108-76-3 | |
C12H7F3O2S | |
GIUAGKGTDDIMHB-UHFFFAOYSA-N | |
4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylic acid | |
2783372 | |
95% |
95% | |
4-phenyl-5-trifluoromethyl thiophene-2-carboxylic acid, 4-phenyl-5-trifluoromethyl-2-thiophenecarboxylic acid, 2-thiophenecarboxylic acid, 4-phenyl-5-trifluoromethyl, 2-thiophenecarboxylicacid, 4-phenyl-5-trifluoromethyl, maybridge3_006560, 4-phenyl-5-trifluoromethyl-thiophene-2-carboxylic acid, 4-phenyl-5-trifluoromethyl-2-thiophene carboxylic acid | |
C1=CC=C(C=C1)C2=C(SC(=C2)C(=O)O)C(F)(F)F | |
272.241 | |
272.25 | |
4-Phenyl-5-(trifluoromethyl)thiophene-2-carboxylic acid |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
4-Phenyl-5-(trifluoromethyl)thiophene-2-carboxylic acid, 95%, Thermo Scientific™