missing translation for 'onlineSavingsMsg'
Learn More

4-Piperidone ethylene ketal, 98+%, Thermo Scientific Chemicals

603.52 SEK - 9194.83 SEK

Chemical Identifiers

CAS 177-11-7
Molecular Formula C7H14NO2
Molecular Weight (g/mol) 144.19
MDL Number MFCD00005976
InChI Key KPKNTUUIEVXMOH-UHFFFAOYSA-O
Synonym 1,4-dioxa-8-azaspiro 4.5 decane, 4-piperidone ethylene ketal, 4-piperidinone ethyl ketal, 4-piperidone-ethylene ketal, 4-piperidone ethylene acetal, 1,4-dioxa-8-azaspiro 4,5 decane, 4-piperidoneethyleneketal, piperidone-4-ethyleneketal, 1,4-dioxa-8-aza-spiro 4.5 decane
PubChem CID 67435
SMILES C1COC2(CC[NH2+]CC2)O1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11409613
View Documents
Thermo Scientific Alfa Aesar
10 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11419613
View Documents
Thermo Scientific Alfa Aesar
50 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11429613
View Documents
Thermo Scientific Alfa Aesar
250 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

4-Piperidone ethylene ketal is used in the synthesis of 1,4-dioxa-8-azaspiro [4,5] deca spirocyclotriphosphazenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Piperidone ethylene ketal is used in the synthesis of 1,4-dioxa-8-azaspiro [4,5] deca spirocyclotriphosphazenes.

Solubility
Soluble in water (Partly miscible).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air.
Specifications

Chemical Identifiers

177-11-7
144.19
KPKNTUUIEVXMOH-UHFFFAOYSA-O
67435
C7H14NO2
MFCD00005976
1,4-dioxa-8-azaspiro 4.5 decane, 4-piperidone ethylene ketal, 4-piperidinone ethyl ketal, 4-piperidone-ethylene ketal, 4-piperidone ethylene acetal, 1,4-dioxa-8-azaspiro 4,5 decane, 4-piperidoneethyleneketal, piperidone-4-ethyleneketal, 1,4-dioxa-8-aza-spiro 4.5 decane
C1COC2(CC[NH2+]CC2)O1

Specifications

177-11-7
99°C to 101°C (14 mmHg)
C7H14NO2
MFCD00005976
Air Sensitive
Soluble in water (Partly miscible).
C1COC2(CC[NH2+]CC2)O1
144.19
143.19
4-Piperidone ethylene ketal
1.117
81°C (177°F)
1.4819
506799
1,4-dioxa-8-azaspiro 4.5 decane, 4-piperidone ethylene ketal, 4-piperidinone ethyl ketal, 4-piperidone-ethylene ketal, 4-piperidone ethylene acetal, 1,4-dioxa-8-azaspiro 4,5 decane, 4-piperidoneethyleneketal, piperidone-4-ethyleneketal, 1,4-dioxa-8-aza-spiro 4.5 decane
KPKNTUUIEVXMOH-UHFFFAOYSA-O
1,4-dioxa-8-azaspiro[4.5]decan-8-ium
67435
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

missing translation for 'einecsNumber' : 205-868-6

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.