missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Thiouridine, ∽98, MP Biomedicals™
Click to view available options
Quantity:
5 mg
250 mg
Specifications
Specifications
Color | Yellow |
Assay Percent Range | ∽98 |
Quantity | 5 mg |
Formula Weight | 260.3 |
Physical Form | Powder |
Chemical Name or Material | 4-Thiouridine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction