missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Trifluoromethyl)diphenyl ether, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
4-(Trifluoromethyl)diphenyl ether is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Storage temperature is 2 - 4°Cand Keep container tightly closed in a dryand well-ventilated place.
4-(Trifluoromethyl)diphenyl ether is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Storage temperature is 2 - 4°Cand Keep container tightly closed in a dryand well-ventilated place.
Specifications
Specifications
Boiling Point | 180°C (10 mmHg) |
Molecular Formula | C6H5FN2O2 |
Refractive Index | 1.5135 |
Quantity | 1 g |
Beilstein | 1974851 |
Synonym | 2-nitro-5-fluoroaniline, 5-fluoro-2-nitrophenylamine, 2-amino-4-fluoronitrobenzene, 3-fluoro-6-nitroaniline, 5-fluoro-2-nitrobenzenamine, benzenamine, 5-fluoro-2-nitro, pubchem4476, acmc-1cegf, 5-fluoro-2-nitro-aniline, intermediates-zcf02632 |
Solubility Information | Slightly soluble in water. |
InChI Key | PEDMFCHWOVJDNW-UHFFFAOYSA-N |
SMILES | C1=CC(=C(C=C1F)N)[N+](=O)[O-] |
IUPAC Name | 5-fluoro-2-nitroaniline |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'