missing translation for 'onlineSavingsMsg'
Learn More

4-Trifluoromethylbenzoic anhydride, 97%, Thermo Scientific Chemicals

799.00 SEK - 2525.00 SEK

Chemical Identifiers

CAS 25753-16-6
Molecular Formula C16H8F6O3
Molecular Weight (g/mol) 362.227
MDL Number MFCD00671577
InChI Key FNAWJOBKLWLHTA-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzoic anhydride, tfba, 4-trifluoromethylbenzoic anhydride, 4-trifluoromethyl benzoyl 4-trifluoromethyl benzoate, benzoic acid,4-trifluoromethyl-, anhydride with 4-trifluoromethyl benzoic acid, acmc-1cg53, p-trifluoromethylbenzoic acid anhydride, bis 4-trifluoromethylbenzoic acid anhydride, benzoic acid, 4-trifluoromethyl-, anhydrideShow MoreShow Less
PubChem CID 2760733
IUPAC Name [4-(trifluoromethyl)benzoyl] 4-(trifluoromethyl)benzoate
SMILES C1=CC(=CC=C1C(=O)OC(=O)C2=CC=C(C=C2)C(F)(F)F)C(F)(F)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15417437
SDS View Documents
Thermo Scientific Alfa Aesar
H34480.03
1 g
799.00 SEK
1g
Please to purchase this item. Need a web account? Register with us today!
15427437
SDS View Documents
Thermo Scientific Alfa Aesar
H34480.06
5 g
2525.00 SEK
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Trifluoromethylbenzoic anhydride, 97%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.