missing translation for 'onlineSavingsMsg'
Learn More

4-Undecanone, 98%, Thermo Scientific™

Product Code. 11480661
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14476-37-0
Molecular Formula C11H22O
Molecular Weight (g/mol) 170.30
MDL Number MFCD00048923
InChI Key NBSLHMOSERBUOV-UHFFFAOYSA-N
Synonym 4-undecanone, heptyl propyl ketone, n-heptyl n-propyl ketone, acmc-1c992
PubChem CID 84467
IUPAC Name undecan-4-one
SMILES CCCCCCCC(=O)CCC

Specifications

Density 0.83
Boiling Point 104°C to 107°C (13 mmHg)
Flash Point 92°C (197°F)
Refractive Index 1.428
Quantity 25 g
Beilstein 1749499
Formula Weight 170.3
Percent Purity 98%
Chemical Name or Material 4-Undecanone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'