Learn More
4-Vinyl-1-cyclohexene, 97%, stabilized, Thermo Scientific Chemicals
236.08 SEK - 668.35 SEK
Chemical Identifiers
CAS | 100-40-3 |
---|---|
Molecular Formula | C8H12 |
Molecular Weight (g/mol) | 108.18 |
MDL Number | MFCD00001576 |
InChI Key | BBDKZWKEPDTENS-UHFFFAOYSA-N |
Synonym | 4-vinylcyclohexene, 4-vinyl-1-cyclohexene, cyclohexene, 4-ethenyl, butadiene dimer, 4-vinylcyclohex-1-ene, 4-vinylcyclohexene-1, cyclohexene, 4-vinyl, cyclohexenylethylene, 4-ethenyl-1-cyclohexene, 1-vinylcyclohexene-3 |
PubChem CID | 7499 |
ChEBI | CHEBI:82377 |
IUPAC Name | 4-ethenylcyclohexene |
SMILES | C=CC1CCC=CC1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10397020
|
Thermo Scientific Acros
140880050 |
5 mL | Glass bottle |
236.08 SEK
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10163570
|
Thermo Scientific Acros
140885000 |
500 mL | Glass bottle |
668.35 SEK
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
100-40-3 | |
108.18 | |
BBDKZWKEPDTENS-UHFFFAOYSA-N | |
7499 | |
4-ethenylcyclohexene |
C8H12 | |
MFCD00001576 | |
4-vinylcyclohexene, 4-vinyl-1-cyclohexene, cyclohexene, 4-ethenyl, butadiene dimer, 4-vinylcyclohex-1-ene, 4-vinylcyclohexene-1, cyclohexene, 4-vinyl, cyclohexenylethylene, 4-ethenyl-1-cyclohexene, 1-vinylcyclohexene-3 | |
CHEBI:82377 | |
C=CC1CCC=CC1 |
Specifications
100-40-3 | |
100.0 | |
Beige-Brown to White | |
126.0°C to 127.0°C | |
Authentic | |
C8H12 | |
1.4625 to 1.4645 | |
05, I, 63 | |
4-vinylcyclohexene, 4-vinyl-1-cyclohexene, cyclohexene, 4-ethenyl, butadiene dimer, 4-vinylcyclohex-1-ene, 4-vinylcyclohexene-1, cyclohexene, 4-vinyl, cyclohexenylethylene, 4-ethenyl-1-cyclohexene, 1-vinylcyclohexene-3 | |
BBDKZWKEPDTENS-UHFFFAOYSA-N | |
4-ethenylcyclohexene | |
108.18 | |
CHEBI:82377 | |
108.18 | |
Powder |
98.5 | |
-101.0°C | |
0.8320g/mL | |
16°C | |
96% min. (GC) | |
C6H9CH=CH2 | |
MFCD00001576 | |
0.832 | |
Solubility in water: 0.05g/L (20°C). Other solubilities: soluble in benzene and ether | |
C=CC1CCC=CC1 | |
25 to 200ppm BHT | |
7499 | |
0.7 mPa.s (20°C) | |
97% | |
4-Vinyl-1-cyclohexene, 99% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Suspected of causing cancer.
Harmful to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
GHS P Statement
Use personal protective equipment as required.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF exposed or concerned: Get medical advice/attention.
IF SWALLOWED: Immediately call a POISON CENT
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-848-9
missing translation for 'rtecsNumber' : GW6650000
missing translation for 'tsca' : TSCA
- 4-Vinyl-1-cyclohexene, stabilized
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Aspiration toxicity Category 1
- Carcinogenicity Category 2
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H351-Suspected of causing cancer.
- H361-Suspected of damaging fertility or the unborn child.
- H412-Harmful to aquatic life with long lasting effects.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P331-Do NOT induce vomiting.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.