Learn More
RETSCH 400 dia. x 65 mmH Stainless Steel Test Sieve, ASTM Certified, Millimeter Pore Sizes
Brand: RETSCH 60.167.001400
4393.75 SEK valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Specifications
1Each | |
Test Sieve | |
Stainless Steel | |
65mm | |
400 mm | |
Stainless Steel |
1.40 mm | |
ASTM E 11 | |
Shaker AS 450 | |
For separation, fractioning, particle size determination | |
Stainless Steel |