Learn More
Schneider Gerd™ 4301 Stainless Steel Double Ended Spoon
Use this spoon for a variety of applications. Schneider Gerd™ 4301 Stainless Steel Double Ended Spoons are ideal for use with a wide range of compounds.
Brand:  Schneider Gerd™ 233
 View more versions of this product
Product Code. 10682902
  • 137.00 SEK / Each
Estimated Shipment: 13-03-2025
to see stock.
Explore more special offers
Description
  • Spoons collect and transport dry substances or liquids
  • Two end shapes offer increased versatility
  • Ideal for working with powders, stirring and mixing
  • Perfect for miniscule amounts of chemicals
Specifications
Specifications
1 Pk.
For Chemical
180 mm
Double Spoon End Spatula
Stainless Steel (4301)
Documents
One moment while we fetch your results.
Product Title
Schneider Gerd™ 4301 Stainless Steel Double Ended Spoon > Length: 180mm