missing translation for 'onlineSavingsMsg'
Learn More

5-[1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl]thiophene-2-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10351995
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 175202-93-4
Molecular Formula C10H7F3N2OS
Molecular Weight (g/mol) 260.234
MDL Number MFCD00052163
InChI Key ITUYPRXQRPMRTL-UHFFFAOYSA-N
Synonym 5-1-methyl-5-trifluoromethyl-1h-pyrazol-3-yl thiophene-2-carbaldehyde, 5-1-methyl-5-trifluoromethyl pyrazol-3-yl thiophene-2-carbaldehyde, 2-1-methyl-5-trifluoromethyl pyrazol-3-yl-thiophene-5-carboxaldehyde, 2-1-methyl-5-trifluoromethyl pyrazol-3-yl thiophene-5-carboxaldehyde, 2-1-methyl-5-trifluoromethyl pyrazol-3-yl thiophene-5-carbaldehyde, 2-thiophenecarboxaldehyde,5-1-methyl-5-trifluoromethyl-1h-pyrazol-3-yl, 5-1-methyl-5-trifluoromethyl-1h-pyrazol-3-yl-2-thiophenecarbaldehyde, 5-1-methyl-5-trifluoromethyl-1h-pyrazol-3-yl-2-thiophenecarboxaldehyde, 5-1-methyl-5-trifluoromethyl-1h-pyrazole-3-yl thiophene-2-carbaldehyde
PubChem CID 2775667
IUPAC Name 5-[1-methyl-5-(trifluoromethyl)pyrazol-3-yl]thiophene-2-carbaldehyde
SMILES CN1C(=CC(=N1)C2=CC=C(S2)C=O)C(F)(F)F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 260.24
Percent Purity 97%
Chemical Name or Material 5-[1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl]thiophene-2-carbaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.