missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(2-Pyridyl)thiophene-2-sulfonyl chloride, Technical Grade, Thermo Scientific™
1220.00 SEK - 4915.00 SEK
Chemical Identifiers
CAS | 151858-64-9 |
---|---|
Molecular Formula | C9H6ClNO2S2 |
Molecular Weight (g/mol) | 259.72 |
MDL Number | MFCD00052284 |
InChI Key | OGOMWPWAEIDEOU-UHFFFAOYSA-N |
Synonym | 5-2-pyridyl thiophene-2-sulfonyl chloride, 5-pyridin-2-yl thiophene-2-sulfonyl chloride, 2-thiophenesulfonylchloride, 5-2-pyridinyl, 5-pyridin-2-yl thiophene-2-sulphonyl chloride, 5-pyridin-2-yl thiophene-2-sulphonyl chloride, tech, acmc-1c5sp, 5-pyrid-2-ylthiophene-2-sulfonyl chloride, chloro 5-2-pyridyl 2-thienyl sulfone, 2-5-chlorosulphonyl thien-2-yl pyridine, 5-2-pyridyl thiophene-2-sulphonyl chloride |
PubChem CID | 2737231 |
IUPAC Name | 5-pyridin-2-ylthiophene-2-sulfonyl chloride |
SMILES | ClS(=O)(=O)C1=CC=C(S1)C1=CC=CC=N1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10742283
|
Thermo Scientific Maybridge
CC02203DA |
1 g |
1220.00 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10776705
|
Thermo Scientific Maybridge
CC02203DE |
5 g |
4915.00 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
151858-64-9 | |
259.72 | |
OGOMWPWAEIDEOU-UHFFFAOYSA-N | |
2737231 | |
ClS(=O)(=O)C1=CC=C(S1)C1=CC=CC=N1 |
C9H6ClNO2S2 | |
MFCD00052284 | |
5-2-pyridyl thiophene-2-sulfonyl chloride, 5-pyridin-2-yl thiophene-2-sulfonyl chloride, 2-thiophenesulfonylchloride, 5-2-pyridinyl, 5-pyridin-2-yl thiophene-2-sulphonyl chloride, 5-pyridin-2-yl thiophene-2-sulphonyl chloride, tech, acmc-1c5sp, 5-pyrid-2-ylthiophene-2-sulfonyl chloride, chloro 5-2-pyridyl 2-thienyl sulfone, 2-5-chlorosulphonyl thien-2-yl pyridine, 5-2-pyridyl thiophene-2-sulphonyl chloride | |
5-pyridin-2-ylthiophene-2-sulfonyl chloride |
Specifications
151858-64-9 | |
C9H6ClNO2S2 | |
5-2-pyridyl thiophene-2-sulfonyl chloride, 5-pyridin-2-yl thiophene-2-sulfonyl chloride, 2-thiophenesulfonylchloride, 5-2-pyridinyl, 5-pyridin-2-yl thiophene-2-sulphonyl chloride, 5-pyridin-2-yl thiophene-2-sulphonyl chloride, tech, acmc-1c5sp, 5-pyrid-2-ylthiophene-2-sulfonyl chloride, chloro 5-2-pyridyl 2-thienyl sulfone, 2-5-chlorosulphonyl thien-2-yl pyridine, 5-2-pyridyl thiophene-2-sulphonyl chloride | |
ClS(=O)(=O)C1=CC=C(S1)C1=CC=CC=N1 | |
259.72 | |
259.74 | |
5-(2-Pyridyl)thiophene-2-sulfonyl chloride |
95% | |
MFCD00052284 | |
OGOMWPWAEIDEOU-UHFFFAOYSA-N | |
5-pyridin-2-ylthiophene-2-sulfonyl chloride | |
2737231 | |
95% |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title