Learn More
5,5-Dimethyl-1,3-dioxane-2-butanal, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
2.5 g
10 g
Description
It is employed as a intermediate for pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as a intermediate for pharmaceuticals.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
It is employed as a intermediate for pharmaceuticals.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Refractive Index | 1.449 |
Quantity | 2.5 g |
Sensitivity | Air Sensitive |
Formula Weight | 186.25 |
Percent Purity | 96% |
Chemical Name or Material | 5,5-Dimethyl-1,3-dioxane-2-butanal |