missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dimethyl-1H-benzotriazole hydrate, 99%, Thermo Scientific Chemicals

572.55 SEK - 2349.49 SEK

Chemical Identifiers

CAS 4184-79-6
Molecular Formula C8H9N3
Molecular Weight (g/mol) 147.18
MDL Number MFCD00149344
InChI Key MVPKIPGHRNIOPT-UHFFFAOYSA-N
Synonym 5,6-dimethyl-1h-benzotriazole hydrate, 5,6-dimethyl-1,2,3-benzotriazole hydrate, 5,6-dimethyl-1h-1,2,3-benzotriazole hydrate, acmc-1aqbb, 1h-benzotriazole,5,6-dimethyl, 5,6-dimethylbenzotriazole, oxamethane, 5,6-dimethyl-2h-benzotriazole hydrate, 5,6-dimethyl-1h-benzotriazole monohydrate, 5,6-dimethyl-1h-benzo d 1,2,3 triazole hydrateShow MoreShow Less
PubChem CID 16219214
IUPAC Name 5,6-dimethyl-2H-benzotriazole;hydrate
SMILES CC1=C(C)C=C2N=NNC2=C1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10366840
SDS View Documents
Thermo Scientific Acros
116050050
5 g Glass bottle N/A N/A N/A
10020850
SDS missing translation for 'viewDocuments'
Thermo Scientific Acros
116050250
25 g Glass bottle
2349.49 SEK
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 224-058-3

Product Identifier
  • 5,6-Dimethyl-1H-benzotriazole hydrate

SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5,6-Dimethyl-1H-benzotriazole hydrate, 99%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.