missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-1-(4-chlorophenyl)-1H-pyrazole-4-carbonitrile, 97%, Thermo Scientific™

200.87 SEK - 1532.05 SEK

Chemical Identifiers

CAS 51516-67-7
Molecular Formula C10H7ClN4
Molecular Weight (g/mol) 218.64
MDL Number MFCD00052031
InChI Key WIWSALMJHPGFDY-UHFFFAOYSA-N
Synonym 5-amino-1-4-chlorophenyl-1h-pyrazole-4-carbonitrile, 5-amino-1-4-chlorophenyl pyrazole-4-carbonitrile, 1h-pyrazole-4-carbonitrile, 5-amino-1-4-chlorophenyl, maybridge1_001852, acmc-1b1ld, 5-amino-1-4-chloro-phenyl-1h-pyrazole-4-carbonit, 1-4-chlorophenyl-5-amino-1h-pyrazole-4-carbonitrileShow MoreShow Less
PubChem CID 639097
SMILES NC1=C(C=NN1C1=CC=C(Cl)C=C1)C#N
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10232392
SDS View Documents
Thermo Scientific Maybridge
BTB07485DA
1 g Amber Glass Bottle
200.87 SEK
1g
Please to purchase this item. Need a web account? Register with us today!
10600903
SDS View Documents
Thermo Scientific Maybridge
BTB07485EA
10 g Amber Glass Bottle
1532.05 SEK
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Amino-1-(4-chlorophenyl)-1H-pyrazole-4-carbonitrile, 97%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.