missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-bromo-3-fluoropyridine, 97%, Thermo Scientific™

Product Code. 15415957
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Commercially available 5-bromo-2-fluoropyridine was treated with LDA at -78°C to give the lithiated species. Fluoropyridine-based bromoacetamide reagent has been prepared and used in prosthetic group introduction for the labeling of oligonucleotides.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Refrigerate(2-7°C ).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1256276-41-1
Molecular Formula C5H4BrFN2
Molecular Weight (g/mol) 191.003
MDL Number MFCD21099397
InChI Key DDJLNVAJTGIWNJ-UHFFFAOYSA-N
Synonym 5-amino-2-bromo-3-fluoropyridine, 3-pyridinamine, 6-bromo-5-fluoro
PubChem CID 71741382
IUPAC Name 6-bromo-5-fluoropyridin-3-amine
SMILES C1=C(C=NC(=C1F)Br)N

Specifications

Melting Point 114°C to 118°C
Quantity 1 g
Formula Weight 191
Percent Purity 97%
Chemical Name or Material 5-Amino-2-bromo-3-fluoropyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'