Learn More
5-Amino-4-bromo-3-methyl-1H-pyrazole hydrobromide, 98%, Thermo Scientific™
279.00 SEK - 3976.00 SEK
Chemical Identifiers
CAS | 167683-86-5 |
---|---|
Molecular Formula | C4H7Br2N3 |
Molecular Weight (g/mol) | 256.929 |
MDL Number | MFCD00053023 |
InChI Key | UGCOULAJNKWOPX-UHFFFAOYSA-N |
Synonym | 5-amino-4-bromo-3-methylpyrazole hydrobromide, 4-bromo-3-methyl-1h-pyrazol-5-amine hydrobromide, 4-bromo-5-methyl-1h-pyrazol-3-amine hydrobromide, 3-methyl-4-bromo-5-aminopyrazole hydrobromide, 4-bromo-5-methyl-2h-pyrazol-3-amine hydrobromide, acmc-20am6c, buttpark 48\04-71, 6-amino-3-bromo-2-methylpyrazole hydrobromide, 3-amino-4-bromo-5-methyl-1h-pyrazole hydrobromide, 5-amino-4-bromo-3-methyl-1h-pyrazole hydrobromide |
PubChem CID | 2778427 |
IUPAC Name | 4-bromo-5-methyl-1H-pyrazol-3-amine;hydrobromide |
SMILES | CC1=C(C(=NN1)N)Br.Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
11421818
|
Thermo Scientific Alfa Aesar
A14487.03 |
1 g | N/A | N/A | N/A | |||||
11431818
|
Thermo Scientific Alfa Aesar
A14487.06 |
5 g | N/A | N/A | N/A | |||||
11441818
|
Thermo Scientific Alfa Aesar
A14487.14 |
25 g | N/A | N/A | N/A | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications5-Amino-4-bromo-3-methyl-1H-pyrazole hydrobromide is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to heat, light and moisture. Handle and store under inert gas.
Chemical Identifiers
167683-86-5 | |
256.929 | |
UGCOULAJNKWOPX-UHFFFAOYSA-N | |
2778427 | |
CC1=C(C(=NN1)N)Br.Br |
C4H7Br2N3 | |
MFCD00053023 | |
5-amino-4-bromo-3-methylpyrazole hydrobromide, 4-bromo-3-methyl-1h-pyrazol-5-amine hydrobromide, 4-bromo-5-methyl-1h-pyrazol-3-amine hydrobromide, 3-methyl-4-bromo-5-aminopyrazole hydrobromide, 4-bromo-5-methyl-2h-pyrazol-3-amine hydrobromide, acmc-20am6c, buttpark 48\04-71, 6-amino-3-bromo-2-methylpyrazole hydrobromide, 3-amino-4-bromo-5-methyl-1h-pyrazole hydrobromide, 5-amino-4-bromo-3-methyl-1h-pyrazole hydrobromide | |
4-bromo-5-methyl-1H-pyrazol-3-amine;hydrobromide |
Specifications
167683-86-5 | |
C4H7Br2N3 | |
5-amino-4-bromo-3-methylpyrazole hydrobromide, 4-bromo-3-methyl-1h-pyrazol-5-amine hydrobromide, 4-bromo-5-methyl-1h-pyrazol-3-amine hydrobromide, 3-methyl-4-bromo-5-aminopyrazole hydrobromide, 4-bromo-5-methyl-2h-pyrazol-3-amine hydrobromide, acmc-20am6c, buttpark 48\04-71, 6-amino-3-bromo-2-methylpyrazole hydrobromide, 3-amino-4-bromo-5-methyl-1h-pyrazole hydrobromide, 5-amino-4-bromo-3-methyl-1h-pyrazole hydrobromide | |
UGCOULAJNKWOPX-UHFFFAOYSA-N | |
4-bromo-5-methyl-1H-pyrazol-3-amine;hydrobromide | |
2778427 | |
98% |
178°C to 181°C | |
MFCD00053023 | |
Soluble in water. | |
CC1=C(C(=NN1)N)Br.Br | |
256.929 | |
256.94 | |
5-Amino-4-bromo-3-methyl-1H-pyrazole hydrobromide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.