missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Aminopyridine-2-carboxylic acid, 98%, Thermo Scientific™
1117.72 SEK - 3375.97 SEK
Chemical Identifiers
CAS | 24242-20-4 |
---|---|
Molecular Formula | C6H6N2O2 |
Molecular Weight (g/mol) | 138.13 |
InChI Key | WDJARUKOMOGTHA-UHFFFAOYSA-N |
Synonym | 5-amino-2-pyridinecarboxylic acid, 5-aminopicolinic acid, 5-amino-2-picolinic acid, 2-pyridinecarboxylic acid, 5-amino, 5-amino-pyridine-2-carboxylic acid, 5-amino-2-pyridinecarboxylicacid, pubchem15896, 5-amino-2-carboxypyridine, acmc-209ga3, ksc201o9n |
PubChem CID | 4179726 |
IUPAC Name | 5-aminopyridine-2-carboxylic acid |
SMILES | C1=CC(=NC=C1N)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15276018
|
Thermo Scientific Acros
450980010 |
1 g |
1117.72 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15286018
|
Thermo Scientific Acros
450980050 |
5 g |
3375.97 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
24242-20-4 | |
138.13 | |
5-amino-2-pyridinecarboxylic acid, 5-aminopicolinic acid, 5-amino-2-picolinic acid, 2-pyridinecarboxylic acid, 5-amino, 5-amino-pyridine-2-carboxylic acid, 5-amino-2-pyridinecarboxylicacid, pubchem15896, 5-amino-2-carboxypyridine, acmc-209ga3, ksc201o9n | |
5-aminopyridine-2-carboxylic acid |
C6H6N2O2 | |
WDJARUKOMOGTHA-UHFFFAOYSA-N | |
4179726 | |
C1=CC(=NC=C1N)C(=O)O |
Specifications
24242-20-4 | |
Authentic | |
Glass Bottle | |
5-amino-2-pyridinecarboxylic acid, 5-aminopicolinic acid, 5-amino-2-picolinic acid, 2-pyridinecarboxylic acid, 5-amino, 5-amino-pyridine-2-carboxylic acid, 5-amino-2-pyridinecarboxylicacid, pubchem15896, 5-amino-2-carboxypyridine, acmc-209ga3, ksc201o9n | |
C1=CC(=NC=C1N)C(=O)O | |
138.13 | |
138.13 | |
5-Aminopyridine-2-carboxylic acid |
215.0°C to 217.0°C | |
97.5 % min. (HPLC) | |
C6H6N2O2 | |
WDJARUKOMOGTHA-UHFFFAOYSA-N | |
5-aminopyridine-2-carboxylic acid | |
4179726 | |
98% |
Safety and Handling
GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 5-Aminopyridine-2-carboxylic acid, 98%
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only