missing translation for 'onlineSavingsMsg'
Learn More

5'-Bromo-2'-hydroxy-3'-nitroacetophenone, 97%, Thermo Scientific Chemicals

Product Code. 11322429
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70978-54-0
Molecular Formula C8H6BrNO4
Molecular Weight (g/mol) 260.04
MDL Number MFCD01631134
InChI Key CLNIBJASCGZXHH-UHFFFAOYSA-N
PubChem CID 689071
IUPAC Name 1-(5-bromo-2-hydroxy-3-nitrophenyl)ethan-1-one
SMILES CC(=O)C1=CC(Br)=CC(=C1O)[N+]([O-])=O

Specifications

Melting Point 129°C to 132°C
Quantity 5 g
Synonym 1-5-bromo-2-hydroxy-3-nitrophenyl ethanone, 5'-bromo-2'-hydroxy-3'-nitroacetophenone, 3-bromo-6-hydroxy-5-nitroacetophenone, 5'-bromo-3'-nitro-2'-hydroxyacetophenone, 5-bromo-2-hydroxy-3-nitroacetophenone, 1-5-bromo-2-hydroxy-3-nitrophenyl ethan-1-one, ethanone,1-5-bromo-2-hydroxy-3-nitrophenyl, ethanone, 1-5-bromo-2-hydroxy-3-nitrophenyl, 3'-bromo-6'-hydroxy-5'-nitroacetophenone, 1-acetyl-5-bromo-2-hydroxy-3-nitrobenzene
Formula Weight 260.05
Percent Purity 97%
Chemical Name or Material 5'-Bromo-2'-hydroxy-3'-nitroacetophenone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'