missing translation for 'onlineSavingsMsg'
Learn More
5-Bromo-3-iodo-2-methoxypyridine, 97%, Thermo Scientific™ Product Code.: 11434460

5-Bromo-3-iodo-2-methoxypyridine, 97%, Thermo Scientific™

Product Code. 11434460
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11434460

Brand: Thermo Scientific Alfa Aesar H50061.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-Bromo-3-iodo-2-methoxypyridine is used as an intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 578007-66-6
Molecular Formula C6H5BrINO
Molecular Weight (g/mol) 313.92
MDL Number MFCD05663522
InChI Key ZKVDYKJRRHTPLT-UHFFFAOYSA-N
Synonym 5-bromo-3-iodo-2-methoxy-pyridine, pubchem8079, acmc-1aulg, pyridine,5-bromo-3-iodo-2-methoxy, 5-bromanyl-3-iodanyl-2-methoxy-pyridine
PubChem CID 1519406
IUPAC Name 5-bromo-3-iodo-2-methoxypyridine
SMILES COC1=NC=C(Br)C=C1I

Specifications

Quantity 1 g
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 313.92
Percent Purity 97%
Chemical Name or Material 5-Bromo-3-iodo-2-methoxypyridine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberI28U030 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Lot NumberI16Q18 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot NumberF06T037 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot NumberK23W037 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot NumberI11T033 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Bromo-3-iodo-2-methoxypyridine, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.