missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-4-Indoyl-α-D-galactopyranoside, >99%, For TLC analysis, MP Biomedicals™
Click to view available options
Quantity:
5 mg
100 mg
Specifications
Specifications
Color | White |
Assay Percent Range | > 99% |
Quantity | 5 mg |
Formula Weight | 408.63 |
Physical Form | Powder |
Chemical Name or Material | 5-Bromo-4-Indoyl-α-D-galactopyranoside, For TLC analysis |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'