missing translation for 'onlineSavingsMsg'
Learn More
5-Bromovaleronitrile, 98+%, Thermo Scientific Chemicals Product Code.: 11366726

5-Bromovaleronitrile, 98+%, Thermo Scientific Chemicals

Product Code. 11366726
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11366726

Brand: Thermo Scientific Alfa Aesar L03952.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

5-Bromovaleronitrile is used to produce 5-Bromo-pentanoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromovaleronitrile is used to produce 5-Bromo-pentanoic acid.

Solubility
Slightly soluble in water (4.1 g/L at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5414-21-1
Molecular Formula C5H8BrN
Molecular Weight (g/mol) 162.03
MDL Number MFCD00001976
InChI Key NWWWGAKVHCSAEU-UHFFFAOYSA-N
Synonym 5-bromovaleronitrile, pentanenitrile, 5-bromo, 5bromovaleronitrile, snxhflqiuzjp@, 5-bromopentanonitrile, 4-bromobutyl cyanide, 5-bromo-valeronitrile, 5-bromo-pentanenitrile, 5-bromopentane nitrile, 1-bromo-4-cyanobutaneShow MoreShow Less
PubChem CID 79435
IUPAC Name 5-bromopentanenitrile
SMILES C(CCBr)CC#N

Specifications

Density 1.388
Boiling Point 110°C to 111°C (12 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4785
Quantity 5 g
UN Number UN3276
Beilstein 1742080
Solubility Information Slightly soluble in water (4.1g/L at 25°C).
Formula Weight 162.04
Percent Purity ≥98%
Chemical Name or Material 5-Bromovaleronitrile
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10177272 Certificate TypeCertificate of Analysis Date11/08/2023 Catalog NumberL03952.06,L03952.14
Lot NumberG09Z028 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL03952.06,L03952.14
Lot Number10117386 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL03952.06,L03952.14
Lot NumberD23Y010 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL03952.06,L03952.14
Lot Number10116846 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL03952.06,L03952.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Bromovaleronitrile, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.