missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-1-pentanol, 95%, Thermo Scientific Chemicals

Product Code. 11398605
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

5-Chloro-1-pentanol is used to produce 6-hydroxy-hexanenitrile at temperature of 90 - 100°C. It is also is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chloro-1-pentanol is used to produce 6-hydroxy-hexanenitrile at temperature of 90 - 100°C. It is also is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5259-98-3
Molecular Formula C5H11ClO
Molecular Weight (g/mol) 122.59
MDL Number MFCD00039556
InChI Key DCBJCKDOZLTTDW-UHFFFAOYSA-N
Synonym 5-chloro-1-pentanol, 5-chloropentanol, pentamethylene chlorohydrin, 1-pentanol, 5-chloro, 1-chloro-5-hydroxypentane, unii-55vp4w35eg, 1-chloro-5-pentanol, 5-chloropentane-1-ol, acmc-1amb4, ksc272o9l
PubChem CID 78915
IUPAC Name 5-chloropentan-1-ol
SMILES OCCCCCCl

Specifications

Density 1.06
Boiling Point 102°C to 103°C (8 mmHg)
Refractive Index 1.454
Quantity 10 g
Beilstein 1732314
Solubility Information Slightly soluble in water.
Formula Weight 122.6
Percent Purity 95%
Chemical Name or Material 5-Chloro-1-pentanol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'