missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chloro-2-methoxybenzonitrile, 98%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is employed as an intermediate for pharmaceutical.
Solubility
Insoluble in water.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
It is employed as an intermediate for pharmaceutical.
Solubility
Insoluble in water.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 98°C to 102°C |
Quantity | 1 g |
UN Number | UN3439 |
Beilstein | 3245801 |
Solubility Information | Insoluble in water. |
Formula Weight | 167.6 |
Percent Purity | 98% |
Chemical Name or Material | 5-Chloro-2-methoxybenzonitrile |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'