missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-(trifluoromethyl)phenylacetonitrile, 97%, Thermo Scientific™

925.00 SEK - 3144.00 SEK

Chemical Identifiers

CAS 261763-26-2
Molecular Formula C9H5ClF3N
Molecular Weight (g/mol) 219.591
MDL Number MFCD01631357
InChI Key XCPSZOAJWIBKNE-UHFFFAOYSA-N
Synonym 5-chloro-2-trifluoromethyl phenylacetonitrile, 2-5-chloro-2-trifluoromethyl phenyl acetonitrile, 5-chloro-2-trifluoromethyl phenyl acetonitrile, 3-chloro-2-6-trifluoromethyl phenylacetonitrile, 2-5-chloro-2-trifluoromethyl phenyl ethanenitrile, pubchem4286, acmc-20am4x, 5-chloro-2-trifluoromethylbenzyl cyanide, 5-chloro-2-trifluoromethyl benzyl cyanide
PubChem CID 2778126
IUPAC Name 2-[5-chloro-2-(trifluoromethyl)phenyl]acetonitrile
SMILES C1=CC(=C(C=C1Cl)CC#N)C(F)(F)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11455667
SDS View Documents
Thermo Scientific Alfa Aesar
B23629.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11465667
SDS View Documents
Thermo Scientific Alfa Aesar
B23629.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

261763-26-2
219.591
XCPSZOAJWIBKNE-UHFFFAOYSA-N
2778126
C1=CC(=C(C=C1Cl)CC#N)C(F)(F)F
C9H5ClF3N
MFCD01631357
5-chloro-2-trifluoromethyl phenylacetonitrile, 2-5-chloro-2-trifluoromethyl phenyl acetonitrile, 5-chloro-2-trifluoromethyl phenyl acetonitrile, 3-chloro-2-6-trifluoromethyl phenylacetonitrile, 2-5-chloro-2-trifluoromethyl phenyl ethanenitrile, pubchem4286, acmc-20am4x, 5-chloro-2-trifluoromethylbenzyl cyanide, 5-chloro-2-trifluoromethyl benzyl cyanide
2-[5-chloro-2-(trifluoromethyl)phenyl]acetonitrile

Specifications

261763-26-2
C9H5ClF3N
UN3439
XCPSZOAJWIBKNE-UHFFFAOYSA-N
2-[5-chloro-2-(trifluoromethyl)phenyl]acetonitrile
2778126
97%
61°C to 63°C
MFCD01631357
5-chloro-2-trifluoromethyl phenylacetonitrile, 2-5-chloro-2-trifluoromethyl phenyl acetonitrile, 5-chloro-2-trifluoromethyl phenyl acetonitrile, 3-chloro-2-6-trifluoromethyl phenylacetonitrile, 2-5-chloro-2-trifluoromethyl phenyl ethanenitrile, pubchem4286, acmc-20am4x, 5-chloro-2-trifluoromethylbenzyl cyanide, 5-chloro-2-trifluoromethyl benzyl cyanide
C1=CC(=C(C=C1Cl)CC#N)C(F)(F)F
219.591
219.59
5-Chloro-2-(trifluoromethyl)phenylacetonitrile
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H312-H332-H315-H319
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H301-H312+H332-H315-H319

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Chloro-2-(trifluoromethyl)phenylacetonitrile, 97%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.