missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Decyne, 98%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
50 g
Description
5-Decyne is used as APIs and intermediates. It is also used for chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Decyne is used as APIs and intermediates. It is also used for chemical research.
Solubility
Not miscible in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
5-Decyne is used as APIs and intermediates. It is also used for chemical research.
Solubility
Not miscible in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Specifications
Density | 0.77 |
Boiling Point | 177°C to 178°C |
Flash Point | 54°C (129°F) |
Odor | Petrol-like |
Refractive Index | 1.433 |
Quantity | 10 g |
UN Number | UN3295 |
Beilstein | 1697840 |
Solubility Information | Not miscible in water. |
Formula Weight | 138.26 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'