Learn More
5-Fluoro-2-methoxyphenylacetonitrile, 97%, Thermo Scientific™
749.08 SEK - 1970.00 SEK
Chemical Identifiers
CAS | 501008-41-9 |
---|---|
Molecular Formula | C9H8FNO |
Molecular Weight (g/mol) | 165.17 |
MDL Number | MFCD00671773 |
InChI Key | BHJFTJPFOVONOM-UHFFFAOYSA-N |
Synonym | 2-methoxy-5-fluorobenzyl cyanide, 2-5-fluoro-2-methoxyphenyl acetonitrile, 5-fluoro-2-methoxyphenylacetonitrile, benzeneacetonitrile,5-fluoro-2-methoxy, 5-fluoro-2-methoxybenzeneacetonitrile, 5-fluoro-2-methoxyphenyl acetonitrile, 5-fluoro-2-methoxybenzyl cyanide, 2-5-fluoro-2-methoxyphenyl ethanenitrile, 2-5-fluoranyl-2-methoxy-phenyl ethanenitrileShow More |
PubChem CID | 302685 |
IUPAC Name | 2-(5-fluoro-2-methoxyphenyl)acetonitrile |
SMILES | COC1=C(CC#N)C=C(F)C=C1 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
501008-41-9 | |
165.17 | |
BHJFTJPFOVONOM-UHFFFAOYSA-N | |
302685 | |
COC1=C(CC#N)C=C(F)C=C1 |
C9H8FNO | |
MFCD00671773 | |
2-methoxy-5-fluorobenzyl cyanide, 2-5-fluoro-2-methoxyphenyl acetonitrile, 5-fluoro-2-methoxyphenylacetonitrile, benzeneacetonitrile,5-fluoro-2-methoxy, 5-fluoro-2-methoxybenzeneacetonitrile, 5-fluoro-2-methoxyphenyl acetonitrile, 5-fluoro-2-methoxybenzyl cyanide, 2-5-fluoro-2-methoxyphenyl ethanenitrile, 2-5-fluoranyl-2-methoxy-phenyl ethanenitrileShow More | |
2-(5-fluoro-2-methoxyphenyl)acetonitrile |
Specifications
501008-41-9 | |
C9H8FNO | |
UN3439 | |
BHJFTJPFOVONOM-UHFFFAOYSA-N | |
2-(5-fluoro-2-methoxyphenyl)acetonitrile | |
302685 | |
97% |
53°C to 56°C | |
MFCD00671773 | |
2-methoxy-5-fluorobenzyl cyanide, 2-5-fluoro-2-methoxyphenyl acetonitrile, 5-fluoro-2-methoxyphenylacetonitrile, benzeneacetonitrile,5-fluoro-2-methoxy, 5-fluoro-2-methoxybenzeneacetonitrile, 5-fluoro-2-methoxyphenyl acetonitrile, 5-fluoro-2-methoxybenzyl cyanide, 2-5-fluoro-2-methoxyphenyl ethanenitrile, 2-5-fluoranyl-2-methoxy-phenyl ethanenitrile | |
COC1=C(CC#N)C=C(F)C=C1 | |
165.17 | |
165.17 | |
5-Fluoro-2-methoxyphenylacetonitrile |
Safety and Handling
GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.