missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-methylphenol, 98%, Thermo Scientific™

Product Code. 11401637
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 452-85-7
Molecular Formula C7H7FO
Molecular Weight (g/mol) 126.13
MDL Number MFCD00673009
InChI Key CKJOSIHYVLHIER-UHFFFAOYSA-N
Synonym 4-fluoro-2-hydroxytoluene, 5-fluoro-o-cresol, 5-fluoro-2-methyl-phenol, pubchem1502, 2-methyl-5-fluorophenol, acmc-1afs5, 5-fluoranyl-2-methyl-phenol, ksc494a2f
PubChem CID 2774616
IUPAC Name 5-fluoro-2-methylphenol
SMILES CC1=C(C=C(C=C1)F)O

Specifications

Density 1.157
Boiling Point 66°C (4 mmHg)
Odor Phenol-like
Refractive Index 1.514
Quantity 1 g
UN Number UN3265
Beilstein 3234863
Formula Weight 126.13
Percent Purity 98%
Chemical Name or Material 5-Fluoro-2-methylphenol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'