missing translation for 'onlineSavingsMsg'
Learn More
5-Fluoro-2-(trifluoromethyl)aniline, 97%, Thermo Scientific™ Product Code.: 11392559

5-Fluoro-2-(trifluoromethyl)aniline, 97%, Thermo Scientific™

Product Code. 11392559
1 g
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy

Product Code. 11392559

Brand: Thermo Scientific Alfa Aesar H26657.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Used to prepare dyes. Industrially used in natural gas and refinery process streams. They also are used in polymer industry as monomers. Amines find use as additives for lubricating oils, engine fuels, and asphalt, and are producers of ethanolamines.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 827-20-3
Molecular Formula C7H5F4N
Molecular Weight (g/mol) 179.118
MDL Number MFCD06660353
InChI Key LBSBTIMHMYAFFS-UHFFFAOYSA-N
Synonym 5-fluoro-2-trifluoromethyl aniline, 2-amino-4-fluorobenzotrifluoride, benzenamine, 5-fluoro-2-trifluoromethyl, pubchem1544, 5-fluoro-2-trifluoromethyl phenylamine
PubChem CID 17750836
IUPAC Name 5-fluoro-2-(trifluoromethyl)aniline
SMILES C1=CC(=C(C=C1F)N)C(F)(F)F

Specifications

Melting Point 29°C to 32°C
Odor Characteristic
Quantity 1 g
UN Number UN2811
Solubility Information Insoluble in water.
Formula Weight 179.12
Percent Purity 97%
Chemical Name or Material 5-Fluoro-2-(trifluoromethyl)aniline

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10181685 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot Number10145515 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot Number10141475 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot Number10154096 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Fluoro-2-(trifluoromethyl)aniline, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.