missing translation for 'onlineSavingsMsg'
Learn More
5-Hexynoic acid, 96%, Thermo Scientific Chemicals Product Code.: 11416187

5-Hexynoic acid, 96%, Thermo Scientific Chemicals

Product Code. 11416187
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11416187

Brand: Thermo Scientific Alfa Aesar B25132.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

5-Hexynoic acid is used in the preparation of hex-5-ynoic acid methyl ester by using p-toluenesulfonic acid as a reagent. It is also involved in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Hexynoic acid is used in the preparation of hex-5-ynoic acid methyl ester by using p-toluenesulfonic acid as a reagent. It is also involved in chemical synthesis.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53293-00-8
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.128
MDL Number MFCD00066346
InChI Key VPFMEXRVUOPYRG-UHFFFAOYSA-N
Synonym 5-hexynoic acid, 5-hexyn-1-oic acid, 4-ethynylbutyric acid, acmc-1an4h
PubChem CID 143036
ChEBI CHEBI:73511
IUPAC Name hex-5-ynoic acid
SMILES C#CCCCC(=O)O

Specifications

Density 0.99
Boiling Point 73°C to 74°C (0.2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4495
Quantity 5 g
UN Number UN3265
Beilstein 1743192
Solubility Information Miscible with water.
Formula Weight 112.13
Percent Purity 97%
Chemical Name or Material 5-Hexynoic acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10205846 Certificate TypeCertificate of Analysis Date10/07/2023 Catalog NumberB25132,B25132.06,B25132.03,B25132.14
Lot Number10201683 Certificate TypeCertificate of Analysis Date06/07/2023 Catalog NumberB25132,B25132.06,B25132.03,B25132.14
Lot NumberT06E016 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberB25132.06,B25132.03,B25132.14
Lot NumberJ12S035 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB25132.06,B25132.03,B25132.14
Lot Number10209826 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB25132.06,B25132.03,B25132.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Hexynoic acid, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.