missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxyindole-3-acetic acid, 98%, Thermo Scientific Chemicals

Metabolite of melatonin

1100.00 SEK - 1785.00 SEK

Chemical Identifiers

CAS 54-16-0
Molecular Formula C10H9NO3
Molecular Weight (g/mol) 191.186
MDL Number MFCD00005639
InChI Key DUUGKQCEGZLZNO-UHFFFAOYSA-N
Synonym 5-hydroxyindole-3-acetic acid, 5-hiaa, 2-5-hydroxy-1h-indol-3-yl acetic acid, 5-hydroxyindoleacetic acid, 5-hydroxyheteroauxin, 5-hydroxy-1h-indol-3-yl acetic acid, 5-oxyindoleacetic acid, hydroxyindoleacetic acid, 5-hydroxy-iaa, 5-hydroxyindol-3-ylacetic acid
PubChem CID 1826
ChEBI CHEBI:27823
IUPAC Name 2-(5-hydroxy-1H-indol-3-yl)acetic acid
SMILES C1=CC2=C(C=C1O)C(=CN2)CC(=O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15404049
SDS View Documents
Thermo Scientific Alfa Aesar
J61435.ME
500 mg
1100.00 SEK
500mg
Please to purchase this item. Need a web account? Register with us today!
15494039
SDS View Documents
Thermo Scientific Alfa Aesar
J61435.03
1 g
1785.00 SEK
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

5-Hydroxyindole-3-acetic acid is a major serotonin metabolite.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Hydroxyindole-3-acetic acid is a major serotonin metabolite.

Solubility
Soluble in ethanol (50 mg/ml).

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 200-195-4

missing translation for 'rtecsNumber' : NL3650000

missing translation for 'tsca' : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Hydroxyindole-3-acetic acid, 98%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.